Anti-LPAR2 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOVA1056-100EA 299 EUR
BIOVA1056-100
Anti-LPAR2 Rabbit Polyclonal Antibody
Antibodies
EDG4 is a member of family I of the G protein-coupled receptors, as well as the EDG family of proteins. This protein functions as a lysophosphatidic acid (LPA) receptor and contributes to Ca2+ mobilization, a critical cellular response to LPA in cells, through association with Gi and Gq proteins. It plays a key role in phospholipase C-beta (PLC-beta) signaling pathway and stimulates phospholipase C (PLC) activity in a manner that is independent of RALA activation.

Type: Primary
Antigen: LPAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human , Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR