Anti-F2 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOVA1114-100EA 299 EUR
BIOVA1114-100
Anti-F2 Rabbit Polyclonal Antibody
Antibodies
Prothrombin is a vitamin K-dependent plasma protein which is synthesized in the liver. Prior to secretion into plasma, prothrombin undergoes post-translational modification by a vitamin K-dependent carboxylase which converts ten specific glutamic acid residues to γ-carboxyglutamic acid (gla). Conversion to thrombin is a key step in the blood coagulation pathway and catalyzes the coagulation of fibrinogen. Clinically, cases of selective deficiency are rare, although, in cases of liver cirrhosis, prothrombin is decreased. During activation, prothrombin is cleaved at Arg271-Thr272 and at Arg320-Ser321 to a "pro" fragment (fragment 1.2) and thrombin, the latter of which is composed of two chains covalently linked by a disulfide bond. There is an additional thrombin feed-back cleavage at Arg284-Thr285 resulting in an additional 13 amino acids being removed from the mature thrombin “A” chain.

Type: Primary
Antigen: F2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR