Anti-Igf1 Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV5121-30TEA 101 EUR
BIOV5121-30T
Anti-Igf1 Rabbit Polyclonal Antibody
IGF-I (insulin-like Growth Factor-I) is a polypeptide growth factor that stimulates the proliferation of a wide range of cell types including muscle, bone, and cartilage tissue. Rat IGF-I is a 7.69 kDa protein.

Type: Primary
Antigen: Igf1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR