Anti-TFRC Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV6963-100EA 299 EUR
BIOV6963-100
Anti-TFRC Rabbit Polyclonal Antibody
Antibodies
Apotransferrin is the ion-free form of Transferrin. Like transferrin, apotransferrin has a physiological role in the transportation and distribution of iron among the body organs. It is also an important transport factor used in defined culture media.

Type: Primary
Antigen: TFRC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR