Anti-Cst3 Mouse Monoclonal Antibody [clone: 2C2]

Supplier: BioVision
BIOVA1023-100EA 333 EUR
BIOVA1023-100
Anti-Cst3 Mouse Monoclonal Antibody [clone: 2C2]
As an inhibitor of cysteine proteinases, this protein is thought to serve an important physiological role as a local regulator of this enzyme activity. Cystatin C is known to inhibit Cathepsin B, H, and L.

Type: Primary
Antigen: Cst3
Clonality: Monoclonal
Clone: 2C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR