Anti-GLB1 Recombinant Monoclonal Antibody [clone: 13R4]

Supplier: BioVision
BIOVA1104-200EA 755 EUR
BIOVA1104-200
Anti-GLB1 Recombinant Monoclonal Antibody [clone: 13R4]
Cleaves beta-linked terminal galactosyl residues from gangliosides, glycoproteins, and glycosaminoglycans. Isoform 2 has no beta-galactosidase catalytic activity, but plays functional roles in the formation of extracellular elastic fibers (elastogenesis) and in the development of connective tissue. Seems to be identical to the elastin-binding protein (EBP), a major component of the non-integrin cell surface receptor expressed on fibroblasts, smooth muscle cells, chondroblasts, leukocytes, and certain cancer cell types. In elastin producing cells, associates with tropoelastin intracellularly and functions as a recycling molecular chaperone which facilitates the secretions of tropoelastin and its assembly into elastic fibers

Type: Primary
Antigen: GLB1
Clonality: Monoclonal
Clone: 13R4
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1, lambda
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR