Anti-PTPRC Mouse Monoclonal Antibody (R-PE) [clone: T200/797]

Supplier: Biotium
BNCR0797-250EA 355 EUR
BNCR0797-250
Anti-PTPRC Mouse Monoclonal Antibody (R-PE) [clone: T200/797]
Recognizes a 180-185 kDa protein, identified as isoform of leukocyte common antigen (CD45RO). This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: T200/797
Conjugation: R-PE
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR