ToxOut™ Endotoxin removal equilibration buffer
Supplier: BioVision
BIOV7940-50EA 141 EUR
BIOV7940-50
ToxOut™ Endotoxin removal equilibration buffer
Buffers Biological Buffers
50 ml of Regeneration buffer for ToxOut Endotoxin Removal Agarose to neutral pH for better detox efficiency and protein recovery.
  • Rapid Endotoxin Removal Agarose
  • High capacity endotoxin removal Agarose, Better Detox
  • Efficiency and protein recovery
Endotoxin is the lipopolysaccharide (LPS) complex located in the outer membrane of gram-negative bacteria. During experimental procedures, large amount of endotoxins are shed and can easily contaminate labware, buffers and downstream products. Presence of endotoxin in any of these has a detrimental effect on culturing of cells and can lead to generation of irreproducible or negative experimental data. Therefore, it is critical to remove endotoxin from samples and products. ToxOutTM Endotoxin Removal Equilibration Buffer is designed to equilibrate BioVision‘s ToxOutTM Rapid Endotoxin Removal Agarose and ToxOutTM High Capacity Endotoxin Removal Agarose to neutral pH for better detox efficiency and protein recovery. Both ToxOutTM Rapid Endotoxin Removal Agarose and ToxOutTM High Capacity Endotoxin Removal Agarose can effectively eliminate endotoxins to ≤0,05 EU/ml in solutions containing proteins or pharmacologically important components.