ToxOut™ High Capacity Endotoxin Removal Regeneration Buffer
Supplier: BioVision
BIOV7945-50EA 141 EUR
BIOV7945-50
ToxOut™ High Capacity Endotoxin Removal Regeneration Buffer
Regenerate ToxOut High Capacity Endotoxin Removal Agarose and spin columns in ToxOutTM High Capacity Endotoxin Removal Kit.
  • Endotoxin-free regeneration buffer
  • Designed for BioVision‘s ToxOut High Capacity Endotoxin Removal Agarose
  • Effectively reduces endotoxins to <0,05EU/ml
Endotoxin is the lipopolysaccharide (LPS) complex located in the outer membrane of gram-negative bacteria. During experimental procedures, large amount of endotoxins are shed and can easily contaminate labware, buffers and downstream products. Therefore, it is critical to remove endotoxin from samples and products. ToxOut High Capacity Endotoxin Removal Regeneration Buffer is the endotoxin-free regeneration buffer containing sodium hydroxide and ethanol. It is specially designed for regenerating BioVision's ToxOut High Capacity Endotoxin Removal Agarose, which effectively reduce endotoxins to <0,05EU/ml in solutions containing proteins or pharmacologically important components.