ToxOut™ High Capacity Endotoxin Removal Wash Buffer

Supplier: BioVision
BIOV7946-50EA 141 EUR
BIOV7946-50
ToxOut™ High Capacity Endotoxin Removal Wash Buffer
Wash ToxOut™ High capacity endotoxin removal agarose and spin columns in ToxOutTM High capacity endotoxin removal kit

  • Endotoxin-free wash buffer
  • Effectively reduce endotoxins to less than 0,05 EU/mL
  • Contains high concentration of salt

Endotoxin is the lipopolysaccharide (LPS) complex located in the outer membrane of gram-negative bacteria. During experimental procedures, large amount of endotoxins are shed and can easily contaminate labware, buffers and downstream products. Therefore, it is critical to remove endotoxin from samples and products. ToxOut High Capacity Endotoxin Removal Wash Buffer is the endotoxin-free wash buffer containing high concentration of salt for washing BioVision’s ToxOut High Capacity Endotoxin Removal Agarose after the regeneration procedure. ToxOutTM High Capacity Endotoxin Removal Agarose can effectively reduce endotoxins to less than 0,05 EU/mL in solutions containing proteins or pharmacologically important components.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR