VWR® VisiLight® CL150, Halogen Cold Light Source
Supplier: VWR
VWR® VisiLight®
631-1772EA 0 EUR
631-1772
VWR® VisiLight® CL150, Halogen Cold Light Source
Lights Cold Light Sources
The VisiLight® halogen cold light source CL150 features modularity and flexibility, and is suitable for a variety of research and industrial applications.
  • Low noise cooling system and vibration-free operation
  • 150 W halogen bulb with 3270 K colour temperature
  • Lightweight, rugged aluminium housing with modern design
  • Variable light intensity from 0 to 100% with electronic dimming and mechanical iris diaphragm
  • Optional filters for daylight (5400 K), red, green, blue and yellow
Ordering information: Supplied without light guides, ring lights, focus optics or filters which must all be ordered separately.