Oil-only sorbent pads
Supplier: Brady
BRDY813747EA 451 EUR
BRDY813747
Oil-only sorbent pads
Bonded oil only sorbent mat with anti-slip backing that absorbs oil and petroleum based liquids. Ultimate sorbent for high volume petroleum spills.
  • Barrier backing limits liquids from penetrating to the floor
  • Can be used indoors and outdoors thanks to the UV resistant top layer
  • Grey colour masks drips and spills for longer use
Heavy duty, needlepunched top layer gives maximum strength and durability, ideal for heavy traffic. Recommended for use in high traffic areas that require high absorption and outstanding durability.
Weight: 9,2 kg