Lockout box, mini
Supplier: Brady
BRDY050938EA 79.8 EUR
BRDY050938
Lockout box, mini
This wall-mountable lockout box is injection moulded using Isoplast polymers for rugged durability.
  • Won't corrode
  • Excellent chemical and heat resistance properties
  • Slot in front allows keys to be inserted when box is locked
  • Transparent PC windows let you view contents
  • Pre-drilled holes in back allow for easy wall-mounting
8 padlocks can be applied. If using cable padlocks, the cable will need to fit through two holes to secure, therefore only 4 padlocks can be applied.
Ordering information: Supplied with 6 hooks for hanging keys, locks, etc. inside the box. Additional hooks have to be ordered separately. Padlocks not included.