MLN-4924 ≥98% (by HPLC)

Supplier: BioVision

Synonyms: Pevonedistat((1S,2S,4R)-4-(4-(((S)-2,3-dihydro-1H-inden-1-yl)amino)-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-2-hydroxycyclopentylmethyl sulfamate

BIOV2566-1000EA 186 EUR
BIOV2566-1000 BIOV2566-500
MLN-4924 ≥98% (by HPLC)
MLN-4924

A potent and selective inhibitor of NEDD8 (Neural precursor cell expressed, developmentally down-regulated 8)-activating enzyme (NAE) (IC50 = 4,7 nM) with potential anticancer activity. NAE activates NEDD8, an ubiquitin-like (UBL) protein that modifies cellular targets in a pathway that is parallel to but distinct from the ubiquitin-proteasome pathway (UPP). Supresses the growth of tumor xenografts in mice.


Soluble in DMSO (~10 mg/ml)

Formula: C₂₁H₂₅N₅O₄S
Storage Temperature: Freezer
CAS Number: 905579-51-3
https://ie.vwr.com/stibo/web/std.lang.all/20/90/23492090.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR