Would you like to log in again?
Handheld refractometers, OPTi
Supplier: BELLINGHAM & STANLEY
OPTi
635-0824EA 396 EUR InStock
635-0824 635-0825 635-0606
Handheld refractometers, OPTi
Refractometers Handheld Refractometers
  • Shallow prism dish made of 316 stainless steel
  • Zero calibration with distilled water
  • Automated temperature compensation (ATC)
Quality instruments for breweries.
Delivery information: Supplied with two batteries, a multi-language guide, a certificate of calibration and a protective case.
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty