Labels for LABXPERT™ and IDXPERT™ printers, type B-482
Supplier: Brady
BRDY804865EA 91.9 EUR
BRDY804865
Labels for LABXPERT™ and IDXPERT™ printers, type B-482
B-482 is designed for tissue cassette identification through processing when used with the Brady BSP™31 Label Attachment System as a supplemental attachment.
It has superior resistance to processing fluids and conditions, retaining legibility including 2D barcoding, and complements the performance of the B-481 StainerBondz™ slide labels when used as part of a complete histology labelling system.