Ionomycin ≥90%
Supplier: MP Biomedicals
|
Warning
|
ICNA0215961101EA
86.6
EUR
ICNA0215961101
Ionomycin ≥90%
Ionomycin
Free Acid
Selective calcium ionophore.
Storage temperature: -20 °C
Formula:
C₄₁H₇₂O₉ |
MDL Number:
MFCD06798385 CAS Number: 56092-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...