Mobile platforms for cleanrooms
TINMCRSTT5H1125EA 3340 EUR
TINMCRSTT5H1125 TINMCRSTT4H900 TINMCRSTT3H675
Mobile platforms for cleanrooms
Easy access platforms with 3, 4 or 5 steps, with feet and casters
  • Ergonomic design
  • Special spring mechanism provides simple handling
  • Solid vibration free handrail allows to maintain balance while climbing or working
  • The perforations allow an 40 % air permeability
  • With non-slip leveling feet and 360 ° rotating casters with brakes
  • Smooth edges prevents tearing and cuts
  • Minimizations of gaps: closed circuit welded joints, preventing accumulation of dust ; easy cleaning
  • Very good resistance against chemicals and all regular cleanroom disinfectants (alcoholic, biocidal and sporicidal)
Made of stainless steel AISI 304 with high 2R gloss polished finish
Cleanroom suitability: ISO class 4, GMP class A.