Precast gels, ExpressPlus™ PAGE gel
Supplier: Genscript
GENSM01115EA 258 EUR
GENSM01115 GENSM41212 GENSM00810 GENSM01215 GENSM81610 GENSM00812 GENSM41215 GENSM42012 GENSM01010 GENSM01012 GENSM01212 GENSM41210 GENSM01210 GENSM42015 GENSM81615 GENSM81612 GENSM42010 GENSM00815
Precast gels, ExpressPlus™ PAGE gel
Electrophoresis Gels Precast Gels
ExpressPlus™ PAGE gels are high performance precast mini PA gels designed for large loading volumes. The unique design of the cassette gives better band resolution and significantly improves the sample distribution in the loading wells which increases the evenness of the band. ExpressPlus™ PAGE Gels are cast in a weak acidic pH buffer that minimises the hydrolysis of polyacrylamide and results in greater gel stability.
Manufactured without SDS, ExpressPlus™ PAGE Gels are ideal for SDS-PAGE and native electrophoresis depending on the running buffer and transfer buffer used. Using specially formulated Tris-MOPS running buffer, ExpressPlus™ PAGE Gels enable proteins to be separated quickly and easily for subsequent detection by staining or Western blotting.
Cassette size: W×L: 84,5×100 mm.