Anti-AR-beta2 Rabbit Polyclonal Antibody [clone: C378]

Supplier: Bioworld Technology
BWRLBS2585EA 283 EUR
BWRLBS2585
Anti-AR-beta2 Rabbit Polyclonal Antibody [clone: C378]
Antibodies
Synthetic peptide, corresponding to amino acids 351-400 of Human AR-beta2.

AR-?2 (C378) pAb detects endogenous levels of AR-?2 protein.

Type: Primary
Antigen: AR-beta2
Clonality: Polyclonal
Clone: C378
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR