Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (22)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Cell Culture Supplements and Additives

https://ie.vwr.com/stibo/smallweb/std.lang.all/99/18/18989918.jpg

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"128"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Yeast extract is the water soluble portion of autolysed yeast containing a source of Vitamin B complex.
Supplier:  VWR Chemicals
Description:   Contains a mixture of protease inhibitors: AEBSF, Aprotinin, E-64, Bestatin and Leupeptin. Each vial can be reconstituted in 1 ml deionised water to f...

Supplier:  VWR Chemicals
Description:   From bovine pancreas.
UOM:  1 * 1 g
Catalog Number: (M306-5ML)

Supplier:  VWR Chemicals
Description:   Contains a mix of protease inhibitors: AEBSF, E-64, Bestatin, EDTA and Pepstatin. Each vial contains sufficient material to provide 5 ml of a 20X solu...
UOM:  1 * 5 ml
Catalog Number: (M250-1ML)

Supplier:  VWR Chemicals
Description:   Contains protease inhibitors for mammalian cell use: AEBSF, Aprotinin, E-64, Bestatin, Leupeptin and Pepstatin
UOM:  1 * 1 ml
Catalog Number: (M307-1ML)

Supplier:  VWR Chemicals
Description:   Contains a mix of protease inhibitors: AEBSF, E-64, Bestatin, Pepstatin, Leupeptin and 1, 10-Phenanthroline. Each vial can be reconstituted in 1 ml di...
UOM:  1 * 1 ml
Supplier:  BIOWEST
Description:   Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and ...
Supplier:  Corning Life Science
Description:   Corning® Matrigel® Matrix, phenol red-free is recommended for assays which require colour detection (i.e. fluorescence). Corning® Matrigel® Basement M...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 8  of 128
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping