AMG-232 ≥98%
Supplier: BioVision
Synonyms: 2-[(3R,5R,6S)-5-(3-chlorophenyl)-6-(4-chlorophenyl)-3-methyl-1-[(2S)-3-methyl-1-propan-2-ylsulfonylbutan-2-yl]-2-oxopiperidin-3-yl]acetic acid
BIOV9556-1EA 90.2 EUR
BIOV9556-1 BIOV9556-5
AMG-232 ≥98%
AMG-232
AMG232 is a highly potent MDM2 inhibitor with remarkable pharmacokinetic properties and in vivo antitumor activity in the SJSA-1 osteosarcoma xenograft model.
  • Crystalline solid soluble in DMSO (>5 mg/ml)
  • Protect from air and light and store at −20 °C
CAS Number: 1352066-68-2