Anti-SERPINA1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BOSSBS-0096R-A555EA 842 EUR
BOSSBS-0096R-A555
Anti-SERPINA1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Alpha-1-antitrypsin is a protease inhibitor, deficiency of which is associated with emphysema and liver disease. The protein is encoded by a gene (PI) located on the distal long arm of chromosome 14.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR