Anti-PKM Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BOSSBS-0101R-A555EA 866 EUR
BOSSBS-0101R-A555
Anti-PKM Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Glycolytic enzyme that catalyzes the transfer of a phosphoryl group from phosphoenolpyruvate (PEP) to ADP, generating ATP. Stimulates POU5F1-mediated transcriptional activation. Plays a general role in caspase independent cell death of tumor cells. The ratio betwween the highly active tetrameric form and nearly inactive dimeric form determines whether glucose carbons are channeled to biosynthetic processes or used for glycolytic ATP production. The transition between the 2 forms contributes to the control of glycolysis and is important for tumor cell proliferation and survival.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PKM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR