Anti-SOX4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BOSSBS-11208R-A350EA 858 EUR
BOSSBS-11208R-A350
Anti-SOX4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SOX4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR