Anti-SPATA4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BOSSBS-11309R-A647EA 858 EUR
BOSSBS-11309R-A647
Anti-SPATA4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
The testis spermatocyte apoptosis-related gene 2 protein (TSARG2, also designated spermatogenesis associated-4 or SPATA4) is involved in spermatogenesis. TSARG2 is specifically expressed in spermatogonia and spermatocytes of the seminiferous tubules, and it localizes to the nucleus. The predicted molecular weight of TSARG2 ranges depending on the species. TSARG2 is significantly upregulated in cryptorchidism and therefore, is a testis-specific apoptosis candidate oncogene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPATA4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR