Anti-EDA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BOSSBS-1149R-HRPEA 866 EUR
BOSSBS-1149R-HRP
Anti-EDA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Seems to be involved in epithelial-mesenchymal signaling during morphogenesis of ectodermal organs. Isoform 1 binds only to the receptor EDAR, while isoform 3 binds exclusively to the receptor XEDAR.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: EDA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR