Anti-SHBG Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BOSSBS-1175R-A350EA 858 EUR
BOSSBS-1175R-A350
Anti-SHBG Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Functions as an androgen transport protein, but may also be involved in receptor mediated processes. Each dimer binds one molecule of steroid. Specific for 5-alpha-dihydrotestosterone, testosterone, and 17-beta-estradiol. Regulates the plasma metabolic clearance rate of steroid hormones by controlling their plasma concentration.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SHBG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR