Anti-GRM2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BOSSBS-1161R-CY5EA 505 EUR
BOSSBS-1161R-CY5
Anti-GRM2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
G-protein coupled receptor for glutamate. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors, such as adenylate cyclase. Signaling inhibits adenylate cyclase activity. May mediate suppression of neurotransmission or may be involved in synaptogenesis or synaptic stabilization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Metabotropic glutamate receptor 2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR