Anti-ADRA2C Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BOSSBS-12040R-A647EA 858 EUR
BOSSBS-12040R-A647
Anti-ADRA2C Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
alpha2C-adrenergic receptors (alpha2C-AR) regulate neurotransmitter release from sympathetic nerves in the heart, and from adrenergic neurons in the central nervous system. alpha2C-AR can influence Parkinson’s disease, panic disorders, and Huntington disease (HD) progression. A genetic variant in the alpha2C-AR coding region (Del322-325) renders the receptor partially uncoupled from Gi, and is a contributing risk factor for heart failure. alpha2C-AR transcripts are present in rat muscle, heart, pancreas, and kidney.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADRA2C
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR