Anti-FERMT2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BOSSBS-12371R-A488EA 858 EUR
BOSSBS-12371R-A488
Anti-FERMT2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Mig-2 recruits migfilin to cell-matrix adhesions, while the interaction with filamin mediates the association of migfilin with actin filaments. Together, Mig-2, migfilin and filamin define a connection between cell matrix adhesions and the actin cytoskeleton and participate in the orchestration of actin assembly and cell shape modulation. Mig-2 expression is transcriptionally elevated in leiomyomas and could be involved in its hormone-mediated growth of leiomyomas of the uterus. Expression of Mig-2 is ubiquitous, and it is found in numerous tumor tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Kindlin 2/MIG2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR