Anti-ZADH2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BOSSBS-13562R-HRPEA 858 EUR
BOSSBS-13562R-HRP
Anti-ZADH2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
ZADH2 belongs to the zinc containing alcohol dehydrogenase family.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ZADH2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR