Anti-TNFRSF9 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BOSSBS-2449R-CY5.5EA 866 EUR
BOSSBS-2449R-CY5.5
Anti-TNFRSF9 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Receptor for TNFSF9/4-1BBL. Possibly active during T cell activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFRSF9
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR