Anti-CD4 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BOSSBS-0766R-CY3EA 866 EUR
BOSSBS-0766R-CY3
Anti-CD4 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Accessory protein for MHC class-II antigen/T-cell receptor interaction. May regulate T-cell activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD4
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR