Anti-HSD17B2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BOSSBS-3856R-A488EA 858 EUR
BOSSBS-3856R-A488
Anti-HSD17B2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone. Also has 20-alpha-HSD activity. Uses NADH while EDH17B3 uses NADPH.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSD17B2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR