Anti-ALDH1A1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BOSSBS-10162R-CY7EA 509 EUR
BOSSBS-10162R-CY7
Anti-ALDH1A1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Binds free retinal and cellular retinol-binding protein-bound retinal. Can convert/oxidize retinaldehyde to retinoic acid (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ALDH1A1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR