Anti-JMJD7-PLA2G4B Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BOSSBS-3780R-CY3EA 505 EUR
BOSSBS-3780R-CY3
Anti-JMJD7-PLA2G4B Rabbit Polyclonal Antibody (Cy3®)
Antibodies
This gene encodes a member of the cytosolic phospholipase A2 protein family. Phospholipase A2 enzymes hydrolyze the sn-2 bond of phospholipids, releasing lysophospholipids and fatty acids. This enzyme may be associated with mitochondria and early endosomes. Most tissues also express read-through transcripts from the upstream gene into this gene, some of which may encode fusion proteins combining the N-terminus of the upstream gene including its JmjC domain with the almost complete coding region of this gene, including the C2 and cytoplasmic phospholipase A2 domains.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JMJD7-PLA2G4B
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR