Anti-KRV-VP1/KRV-VP2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BOSSBS-4642R-CY5EA 866 EUR
BOSSBS-4642R-CY5
Anti-KRV-VP1/KRV-VP2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Kilham Rat Virus/KRV-VP1/KRV-VP2 (C-terminus)

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KRV-VP1/KRV-VP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR