Anti-NDV HN Protein Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BOSSBS-4529R-CY3EA 509 EUR
BOSSBS-4529R-CY3
Anti-NDV HN Protein Rabbit Polyclonal Antibody (Cy3®)
Antibodies
NDV HN protein

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NDV HN protein
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR