Anti-ELK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BOSSBS-5336R-A555EA 886 EUR
BOSSBS-5336R-A555
Anti-ELK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
This gene is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily. Proteins of the TCF subfamily form a ternary complex by binding to the the serum response factor and the serum response element in the promoter of the c-fos proto-oncogene. The protein encoded by this gene is a nuclear target for the ras-raf-MAPK signaling cascade. This gene produces multiple isoforms by using alternative translational start codons and by alternative splicing. Related pseudogenes have been identified on chromosomes 7 and 14. [provided by RefSeq, Mar 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ELK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR