Anti-SNX27 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BOSSBS-6424R-CY3EA 505 EUR
BOSSBS-6424R-CY3
Anti-SNX27 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
SNX27 is involved in endocytic trafficking. In T lymphocytes it participates in the endocytic recycling pathway. SNX27 recruits PSCDBP and HT4R to early endosomes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SNX27
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR