Anti-CNNM2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BOSSBS-6912R-CY3EA 505 EUR
BOSSBS-6912R-CY3
Anti-CNNM2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Divalent metal cation transporter. Mediates transport of divalent metal cations in an order of Mg(2+) >Co(2+) >Mn(2+) >Sr(2+) >Ba(2+) >Cu(2+) >Fe(2+) (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNNM2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR