Anti-SEPT10 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BOSSBS-7770R-CY5EA 858 EUR
BOSSBS-7770R-CY5
Anti-SEPT10 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
The SEPT10 gene encodes a member of the septin family of cytoskeletal proteins which have GTPase activity. This protein localizes to the cytoplasm and nucleus and displays GTP-binding and GTPase activity. Alternate splicing results in two transcript variants encoding different isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SEPT10
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR