Anti-CDCA8 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BOSSBS-7834R-CY5.5EA 858 EUR
BOSSBS-7834R-CY5.5
Anti-CDCA8 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Component of the chromosomal passenger complex (CPC), a complex that acts as a key regulator of mitosis. The CPC complex has essential functions at the centromere in ensuring correct chromosome alignment and segregation and is required for chromatin-induced microtubule stabilization and spindle assembly. In the complex, it may be required to direct the CPC to centromeric DNA. Major effector of the TTK kinase in the control of attachment-error-correction and chromosome alignment.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDCA8
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR