Anti-KBTBD4 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BOSSBS-8045R-CY5EA 858 EUR
BOSSBS-8045R-CY5
Anti-KBTBD4 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
KBTBD4

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KBTBD4
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR