Anti-COA4 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BOSSBS-8148R-CY5EA 858 EUR
BOSSBS-8148R-CY5
Anti-COA4 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
CHCHD8

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COA4
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR