Anti-BRIX1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BOSSBS-8436R-A350EA 505 EUR
BOSSBS-8436R-A350
Anti-BRIX1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
BXDC2 is a 306 amino acid protein encoded by the human gene BXDC2. BXDC2 is a nuclear protein that contains one Brix domain. Brix domain containing proteins represent a family of proteins involved in the biogenesis of large ribosomal subunits. The Brix domain is a region that is homologous to the yeast protein Pitx1 (Ribosome biogenesis protein BRX1). Pitx1 is part of a complex that includes BXDC5, BXDC1 and PPAN. This complex is required for the biogenesis of the 60S ribosomal subunit. BXDC2 exhibits the same functions as Pitx1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BRIX1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR