Anti-BARX1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BOSSBS-8521R-FITCEA 505 EUR
BOSSBS-8521R-FITC
Anti-BARX1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
BarX1 belongs to the Bar subclass of the homeobox gene family. The function of this gene has not yet been determined; however, studies in the mouse and chick homolog suggest a role in developing teeth and craniofacial mesenchyme of neural crest origin. The role of these homologs implicates the human gene as a candidate for unmapped disorders involving tooth and jaw development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BARX1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR