Anti-CCDC152 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BOSSBS-8619R-CY5.5EA 505 EUR
BOSSBS-8619R-CY5.5
Anti-CCDC152 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
CCDC152

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC152
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR