Anti-IQCG Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BOSSBS-9022R-A555EA 505 EUR
BOSSBS-9022R-A555
Anti-IQCG Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
IQCG is a 443 amino acid protein containing one IQ domain. Widely distributed in nature, the IQ domain forms an amphiphilic seven-turn α-helix capable of binding calmodulin in a Ca2+-independent manner. The level of intracellular calcium is tightly regulated in all eukaryotic cells. A modest increase in this level can result in a myriad of physiological responses, most of which are mediated by calmodulin (CaM), the universal calcium sensor. In acute T-lymphoid/myeloid leukemia, IQCG forms a complex with Nup98, an O-linked glycoprotein and a component of the nuclear pore complex. NUP98-IQCG complex bind co-activators and/or co-repressors, which suggest a role in transcriptional regulation.Nup98-IQCG complex inhibits 32Dcl3 cell apoptosis induced by Arabinofuranosylcytosine (Ara-C) and partially blocks granulocyte differentiation induced by G-CSF. IQCG exists as two isoforms due to alternatively splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IQCG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR